missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 4-1BB/TNFRSF9/CD137 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
£168.00 - £1810.00
Specifications
| Antigen | 4-1BB/TNFRSF9/CD137 |
|---|---|
| Clone | 158321 |
| Dilution | Agonist Activity |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705689
![]() |
R&D Systems
MAB9371-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797398
![]() |
R&D Systems
MAB9371-100 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18745309
![]() |
R&D Systems
MAB9371-500 |
500 μg |
£1810.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Agonist Activity.Specifications
| 4-1BB/TNFRSF9/CD137 | |
| Agonist Activity | |
| Unconjugated | |
| Rat | |
| Agonist Activity | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 41BB, 4-1BB, 4-1BB ligand receptor, CD137, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILA, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, TNFRSF9, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| TNFRSF9 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 158321 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P20334 | |
| 3604 | |
| Mouse myeloma cell line NS0-derived recombinant mouse 4-1BB Val24-Leu187 Accession # P20334 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse 4-1BB in direct ELISAs and Western blots. In direct ELISAs, this antibody shows no cross-reactivity with recombinant mouse (rm) BAFF R, rmCD27, rmCD30, rmCD40, rmDR3, rmEDAR, rmFas, rmGITR, rmLT beta R, rmNGF R, rmOPG, rmRANK, rmTAJ, rmTNF RI, rmTNF RII, rh4-1BB, rhDR6, or rhHVEM. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title