missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse 4-1BB/TNFRSF9/CD137 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB9371-100
This item is not returnable.
View return policy
Description
4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Agonist Activity.
Specifications
| 4-1BB/TNFRSF9/CD137 | |
| 158321 | |
| Agonist Activity | |
| P20334 | |
| TNFRSF9 | |
| Mouse myeloma cell line NS0-derived recombinant mouse 4-1BB Val24-Leu187 Accession # P20334 | |
| 100 μg | |
| Agonist Activity | |
| 3604 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 41BB, 4-1BB, 4-1BB ligand receptor, CD137, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILA, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, TNFRSF9, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Detects mouse 4-1BB in direct ELISAs and Western blots. In direct ELISAs, this antibody shows no cross-reactivity with recombinant mouse (rm) BAFF R, rmCD27, rmCD30, rmCD40, rmDR3, rmEDAR, rmFas, rmGITR, rmLT beta R, rmNGF R, rmOPG, rmRANK, rmTAJ, rmTNF RI, rmTNF RII, rh4-1BB, rhDR6, or rhHVEM. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction