missing translation for 'onlineSavingsMsg'
Learn More

Morin, MP Biomedicals™

Product Code. 11494921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494921 5 g 5g
11404931 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494921 Supplier MP Biomedicals Supplier No. 0215573305
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 480-16-0
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.238
InChI Key YXOLAZRVSSWPPT-UHFFFAOYSA-N
Synonym morin, aurantica, calico yellow, al-morin, toxylon pomiferum, bois d,arc, morin hydrate, 2',3,4',5,7-pentahydroxyflavone, osage orange, osage orange extract
PubChem CID 5281670
ChEBI CHEBI:75092
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one
SMILES C1=CC(=C(C=C1O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O

Specifications

Color Yellow to Brown
Quantity 5 g
Formula Weight 302.2
Physical Form Powder
Chemical Name or Material Morin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.