missing translation for 'onlineSavingsMsg'
Learn More

Morin hydrate

Product Code. p-170121 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
This item is not returnable. View return policy

Product Code. 10589643

Brand: Thermo Scientific Acros 354000050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 654055-01-3 | C15H10O7 | 302.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654055-01-3, 480-16-0
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.24
MDL Number MFCD00217054
InChI Key YXOLAZRVSSWPPT-UHFFFAOYSA-N
Synonym morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
PubChem CID 16219651
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
SMILES OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 300.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 18, 239
Merck Index 14, 6269
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water, soluble in organic solvents
Formula Weight 302.24
Physical Form Powder
Chemical Name or Material Morin hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Morin hydrate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.