Learn More
Merck Montage™ SEQ96 Sequencing Reaction Cleanup Kit
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Brand:  Merck LSKS09604
Code : 71
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11824041
  • £695.00 / Pack of 4
Estimated Shipment: 06-06-2024
to see stock.
Description
Description
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
Specifications
Specifications
DNA sequencing
96-well filter plates and Injection solution
4 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products