Learn More
Merck Montage™ SEQ96 Sequencing Reaction Cleanup Kit
Use 96-well filter plates with proprietary size-exclusion membranes to retain sequencing products while excess dye terminator is removed quickly and efficiently. Fully automatable kits rinse away unincorporated dye terminator without the need for binding and eluting protocols.
£695.00 - £3400.00
Specifications
For Use With (Application) | DNA sequencing |
---|---|
Includes | 96-well filter plates and Injection solution |
Includes: Clear polystyrene 96-well plates, optimized injection solution
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12390180
|
Merck
LSKS09624 |
24 Pack |
£3400.00
Pack of 24 |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11824041
|
Merck
LSKS09604 |
4 Pack |
£695.00
Pack of 4 |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Recovered sequencing products are highly purified and result in long, accurate sequencing reads
- Protocols are optimized for use with BigDye™ Terminator version 3.0, 3.1, 1.1 and ET Terminator sequencing chemistries
- Sequencing products may be resuspended in any aqueous electrokinetic injection buffer
- Plates are suitable for use with 1/8 sequencing reactions, reducing reagent costs by miniaturization
Specifications
DNA sequencing | |
96-well filter plates and Injection solution |