missing translation for 'onlineSavingsMsg'
Learn More

Monobutyl Phthalate, TRC

Product Code. 30008911
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008911 50 mg 50mg
30010220 100 mg 100mg
30012322 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008911 Supplier Toronto Research Chemicals Supplier No. TRCM52510050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-70-4
Molecular Formula C12 H14 O4
Molecular Weight (g/mol) 222.24
Synonym 1,2-Benzenedicarboxylic acid, 1-butyl ester, 1,2-Benzenedicarboxylic acid, monobutyl ester (9CI), Phthalic acid, butyl ester (6CI,7CI), Phthalic acid, monobutyl ester (8CI), Butyl hydrogen phthalate, Mono-n-butyl phthalate, Monobutyl phthalate, NSC 8479, Phthalic acid monobutyl ester
IUPAC Name 2-butoxycarbonylbenzoic acid
SMILES CCCCOC(=O)c1ccccc1C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H14O4/c1-2-3-8-16-12(15)10-7-5-4-6-9(10)11(13)14/h4-7H,2-3,8H2,1H3,(H,13,14)
Formula Weight 222.0892
Percent Purity >95%
Chemical Name or Material Phthalic Acid Monobutyl Ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.