CAS RN 131-70-4
CAS RN 131-70-4
IUPAC Name:
2-(butoxycarbonyl)benzoic acid
Synonyms:
monobutyl phthalate
Molecular Weight (g/mol):
222.24
Molecular Formula:
C12H14O4
InChi Key:
YZBOVSFWWNVKRJ-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C1=CC=CC=C1C(O)=O
1
–
2
of
2
results
1
Phthalic acid, mono-n-butyl ester 100 μg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Monobutyl Phthalate, TRC
CAS: 131-70-4 Molecular Formula: C12 H14 O4 Molecular Weight (g/mol): 222.24 Synonym: 1,2-Benzenedicarboxylic acid, 1-butyl ester,1,2-Benzenedicarboxylic acid, monobutyl ester (9CI),Phthalic acid, butyl ester (6CI,7CI),Phthalic acid, monobutyl ester (8CI),Butyl hydrogen phthalate,Mono-n-butyl phthalate,Monobutyl phthalate,NSC 8479,Phthalic acid monobutyl ester IUPAC Name: 2-butoxycarbonylbenzoic acid SMILES: CCCCOC(=O)c1ccccc1C(=O)O