missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Mono S PC, Columns Prepacked with MonoBeads™
Shop All Cytiva Products
Click to view available options
Height (Metric):
50 mm
100 mm
Max. Pressure:
40 bar
50 bar
Material:
Borosilicate Glass
Glass
PEEK
Unit Size:
Each
Description
- Maximum resolution achieved from efficiency of small spherical particles, optimally packed
- Particles are coupled to Q or S ion exchangers with excellent sensitivities
- Compatible with high-performance LC systems
- Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid
- Tricorn columns are optimized for the polishing step when high purity is required
- PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
- Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
- Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL
Specifications
Specifications
| Buffer | All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid |
| Product Type | Mono S ion exchange column |
| Bed Volume | 1 mL |
| Diameter (English) Inner | 0.19 in. |
| Diameter (Metric) Inner | 5 mm |
| Quantity | Each |
| Height (English) | 1.96 in. |
| Height (Metric) | 50 mm |
| Flow Rate | 0.05 to 3mL/min. |
| Material | Borosilicate Glass |
| Show More |
Safety and Handling
Product Identifier
- Mono S 5/50 GL
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction