missing translation for 'onlineSavingsMsg'
Learn More

MON1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen MON1A
Concentration 0.59 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16802445
View Documents
Proteintech
23772-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16892435
View Documents
Proteintech
23772-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.
Specifications

Specifications

MON1A
Western Blot
Unconjugated
Rabbit
Human
Q86VX9
84315
MON1A Fusion Protein Ag6900
Primary
-20°C
MON1A
0.59 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
MON1 homolog A (yeast), MON1A, SAND1
MON1A
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.