missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MON1A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23772-1-AP-150UL
This item is not returnable.
View return policy
Description
MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.Specifications
| MON1A | |
| Polyclonal | |
| Unconjugated | |
| MON1A | |
| MON1 homolog A (yeast), MON1A, SAND1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84315 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86VX9 | |
| MON1A | |
| MON1A Fusion Protein Ag6900 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction