missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OPP (O-propargyl-puromycin)

Product Code. 15451094
Click to view available options
Quantity:
5 mg
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 15451094

Brand: Invitrogen™ C10459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alkyne analog of puromycin

OPP (O-propargyl-puromycin) is an alkyne analog of puromycin that is efficiently incorporated into newly translated proteins in complete methionine-containing media. When used with Click-iT® Plus OPP Alexa Fluor® Protein Synthesis assays or Alexa Fluor® Picolyl Azide toolkits, OPP provides a fast, sensitive, and non-radioactive method for the detection of protein synthesis using fluorescence microscopy, high-content imaging, or flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and desiccate.
For Use With (Application) Fluorescence Microscopy, High-content Imaging, Flow Cytometry
Product Type OPP
Product Line OPP (O-propargyl-puromycin)
Quantity 5 mg
Reagent Type Nascent Protein Labeling Reagent

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.