missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast Mitochondrial Stain Sampler Kit

Product Code. 10695773
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10695773

Brand: Invitrogen™ Y7530

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Cell-permeant probes found to label mitochondria in live yeast

The Yeast Mitochondrial Stain Sampler Kit contains sample quantities of five different probes including: well characterized mitochondrial stains rhodamine 123, and DiOC6(3), rhodamine B hexyl ester, MitoTracker™ Green FM and SYTO™ 18 yeast mitochondrial stain, a proprietary green fluorescent nucleic acid stain. Each of these probes is sequestered in functioning mitochondria.

Analysis of Yeast, Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbial Tracking, Microbiological Analysis, Mitochondria, Yeast Subcellular Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 1 kit
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Yeast Mitochondrial Stain Sampler Kit
SubCellular Localization Mitochondria

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.