Learn More
Invitrogen™ Vybrant™ Multidrug Resistance Assay Kit
Description
This assay uses the fluorogenic dye calcein, AM as a substrate for efflux activity of Pgp. Calcein AM is nonfluorescent, highly lipid-soluble dye that can rapidly penetrate the plasma membrane of normal cells. Once inside the cell, ester bonds are cleaved by endogenous esterase, transforming calcein AM into the hydrophilic and intensely fluorescent dye, calcein.
- Format: 96-well plate
- Platform: Microplate Reader
- Detection Method: Fluorescent
Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, High-Content Screening (HCS)
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Multidrug Resistance Assay |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Stain |
| Dye Type | Other Label(s) or Dye(s) |
| Format | 96-well plate |
| Product Line | Vybrant |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.