missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ Cell Adhesion Assay Kit

Product Code. 10308262
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10308262 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10308262 Supplier Invitrogen™ Supplier No. V13181

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Sensitive assay for measuring cell-cell or cell-surface adhesion

The Vybrant™ Cell Adhesion Assay Kit is a fast and sensitive assay for measuring cell-cell or cell-surface adhesion for a variety of cell types. In this assay, cells are labeled with calcein AM and allowed to adhere. After removal of nonadherent cells, calcein fluorescence is used to calculate the number of adherent cells.

Cell Adhesion, Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, General Cell Tracing, High-Content Screening (HCS), Neuronal Tracing, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Adhesion Assay
For Use With (Equipment) Microplate Reader
Product Type Stain
Dye Type Other Label(s) or Dye(s)
Format 96-well plate
Product Line Vybrant
Quantity 1 kit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.