missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Transferrin From Human Serum, Alexa Fluor™ 647 Conjugate

Product Code. 11550766
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity unitSize
11550766 5 mg 5mg
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11550766

Brand: Invitrogen™ T23366

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Labeled LDL complexes useful in studying the delivery of up to two Fe3+ atoms via receptor-mediated endocytosis

Transferrin is a monomeric serum glycoprotein (approx. 80,000 daltons) that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis.

  • Once iron-carrying transferrin proteins are inside endosomes, acidic environment favors dissociation of iron from transferrin-receptor complex
  • Following release of iron, apotransferrin is recycled to plasma membrane, where it is released from its receptor to scavenge more iron
  • Fluorescent transferrin conjugates can therefore be used with fluorescent LDL to distinguish lysosomally directed and recycling endosomal pathways
  • These experiments are typically performed by adding fluorescently labeled transferrin to cultured cells and analyzing them by microscopy
  • Label (Ex/Em): Alexa Fluor 594 (590/617)
  • Amount: 15mg solid (contains 5mg of transferrin conjugate)
  • Applications include: Image transferrin receptor dynamics using FRET; Observe receptor trafficking in live cells by confocal laser-scanning microscopy; Investigate events occurring during endosomal acidification; Measure transferrin receptor binding affinity in mammals and parasites

Analysis of Bacteria, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, LPS Binding, Microbiological Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Transferrin
Dye Type Alexa Fluor Dyes
Form Solid
pH 7.2
Product Line Alexa Fluor
Quantity 5 mg
Shipping Condition Room Temperature
Excitation/Emission 650/665
Protein Family Transferrin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.