missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sulforhodamine B

Product Code. 11520666
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11520666

Brand: Invitrogen™ S1307

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Water-soluble, nonfixable red fluorescent (excitation/emission maxima approximately 565/586nm) polar tracer

  • Can be used for investigating neuronal morphology, preparing fluorescent liposomes, studying cell-cell communications and following phagosome-lysosome uptake and fusion

Cell Analysis, Cell Tracing and Tracking, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 5 g
Detection Method Fluorescence
Shipping Condition Room Temperature
Product Type Sulforhodamine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.