missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sulforhodamine 101

Product Code. 11570676
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11570676 25 mg Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11570676 Supplier Invitrogen™ Supplier No. S359
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Water-soluble, nonfixable red fluorescent (excitation/emission maxima approximately 586/605nm) polar tracer

  • Can be used for investigating neuronal morphology, preparing fluorescent liposomes, studying cell-cell communications and following phagosome-lysosome uptake and fusion

Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, General Cell Tracing, Neuronal Tracing

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 25 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Product Type Sulforhodamine

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.