Learn More
Invitrogen™ SP-DiOC18(3) (3,3'-Dioctadecyl-5,5'-Di(4-Sulfophenyl)Oxacarbocyanine, Sodium Salt)
Description
The green fluorescent, lipophilic carbocyanine SP-DiOC18(3) is a DiO analog that contains sulfonate groups to improve water solubility. It is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes. The sulfonate groups incorporated into this DiI analog improves water solubility. It has an extremely high extinction coefficient and short excited-state lifetimes (∽1 nanosecond) in lipid environments.
Cell Analysis, Cell Tracing & Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Green |
| Content And Storage | Store at room temperature and protect from light. |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 5 mg |
| Detection Method | Fluorescence |
| Shipping Condition | Room Temperature |
| Label Type | Fluorescent Dye |
| Product Type | Liphophilic Tracer |
| SubCellular Localization | Plasma Membrane |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.