missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rhodamine B, Hexyl Ester, Perchlorate (R 6)

Product Code. 11533697
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
11533697 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 11533697

Brand: Invitrogen™ R648MP

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cell-permeant, cationic, orange-fluorescent dye

  • Rhodamine B, hexyl ester, perchlorate (R 6) is readily sequestered by active mitochondria with excitation/emission maxima ∽528/551nm
  • Each is for a 10mg size

Cell Analysis, Cell Structure, Mitochondria

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Rhodamine B Hexyl Ester Perchlorate (R 6)
Quantity 10 mg
Shipping Condition Room Temperature
Sub Cellular Localization Mitochondria

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.