Learn More
Invitrogen™ Rhod-2, AM, cell permeant
Description
Labeled calcium indicators are molecules that exhibit increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring Ca2+ in cells and tissues that have high levels of autofluorescence and also for detecting Ca2+ release generated by photoreceptors and photoactivatable chelators.
- Fluorescence signal from these cells is generally measured using fluorescence microscopy
- Label (Ex/Em of Ca2+-bound form): Rhod-2 (552/581nm)
- Fluorescence intensity increase upon binding Ca2+: >100 fold
- Kd for Ca2+ in the absence of Mg2+, in buffer: approximately 570nM
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
- Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Stain |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 20 x 50 μg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.