missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RH 795
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
One of an extensive series of dialkylaminophenylpolyenylpyridinium dyes, RH 795 has produced negligible side effects when tested in vitro using hippocampal slices and in vivo using single-unit recordings in cat and monkey visual cortices.
Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Red |
| Detection Method | Fluorescence |
| Product Type | RH 795 Dye |
| Quantity | 1 mg |
| Content And Storage | Store at room temperature and protect from light. |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Fluorescence Microscope |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction