missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RH 237 (N-(4-Sulfobutyl)-4-(6-(4-(Dibutylamino)phenyl)hexatrienyl)Pyridinium, Inner Salt)

Product Code. 11570656
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11570656 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11570656 Supplier Invitrogen™ Supplier No. S1109
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fast-response potential-sensitive probe

RH 237 can be used for imaging functional neurons.

Cell Analysis, Cell Metabolism, Cell Structure, Cell Viability, Proliferation & Function, Membranes (General) & Lipids

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Infrared
Detection Method Fluorescence
Product Type Sensitive Probe
Quantity 5 mg
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.