missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OxyBURST™ Green H2DCFDA, SE

Product Code. 11510286
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11510286 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11510286 Supplier Invitrogen™ Supplier No. D2935
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Amine-reactive assay reagent

OxyBURST™ Green H2DCFDA succinimidyl ester (2',7'-dichlorodihydrofluorescein diacetate, SE) is an amine-reactive assay reagent that can be used to prepare oxidation-sensitive conjugates of a wide variety of biomolecules and particles, including antibodies, antigens, peptides, proteins, dextrans, bacteria, yeast and polystyrene microspheres. Following conjugation to amines, the two acetates of OxyBURST™ Green H2DCFDA can be removed by treatment with hydroxylamine at neutral pH to yield the dihydrofluorescein conjugate. OxyBURST™ Green H2DCFDA conjugates are nonfluorescent until they are oxidized to the corresponding fluorescein derivatives.

ROS Indicator Specifications:

  • Ex/Em: ∼492–495/517–527 nm
  • Product is air sensitive and should be stored under dry argon or nitrogen
  • Product may be dissolved in DMSO, DMF, or ethanol for use
  • Indicator is cell permeant (cell loading protocols are available in the literature)
  • Fluorescence can be monitored using a flow cytometer, fluorometer, microplate reader, or fluorescence microscope, using excitation sources and filters appropriate for fluorescein

Find More ROS Indicators

We offer an assortment of Molecular Probes™ products for the generation of reactive oxygen species (ROS), including singlet oxygen, superoxide, hydroxyl radical and various peroxide and hydroperoxides, as well as for their fluorometric detection in solution. Review Generating and Detecting Reactive Oxygen Species—Section 18.2 in the Molecular Probes™ Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
For Use With (Application) ROS
Detection Method Fluorescence
Product Type ROS Indicator
Excitation Wavelength Range 492 to 495 nm
Emission Wavelength Range 517 to 527 nm
compliance-icons compliance-icons
Product Identifier
  • 2',7'-DICHLORODIHYDROFLUO
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2'',7''-DICHLORODIHYDROFLUO

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.