missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oregon Green™ 488 BAPTA-1 dextran, Potassium Salt, 10,000 MW, Anionic

Product Code. 11579186
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11579186 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11579186 Supplier Invitrogen™ Supplier No. O6798
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dextran forms of these calcium indicators show dramatic reduction in both leakage and compartmentalization compared to AM ester forms

Labeled calcium indicators are molecules that exhibit increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues.

  • Cells may be physically loaded with cell-impermeant salt forms of these dextran-conjugated indicators using patch pipet or microinjection
  • Fluorescence signal from these cells is measured using fluorescence microscopy
  • Label (Ex/Em): Oregon Green 488 BAPTA-1 (494/523nm)
  • Fluorescence intensity increase upon binding Ca2+: approximately 14 fold
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
  • These probes are excited by visible light, and because energy required for excitation is low, potential for cellular photodamage is reduced
  • Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of spectrum where cellular autofluorescence and scattering backgrounds are often less of problem

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Potassium Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product Type Reagent
Dye Type Fluorescent Dye-Based
Product Line Oregon Green
Quantity 5 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.