missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucRed™ Dead 647 ReadyProbes™ Reagent (TO-PRO-3 iodide)

Product Code. 15139325
Click to view available options
Quantity:
6 Bottles kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15139325

Brand: Invitrogen™ R37113

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

6 x 2.5mL dropper bottles

Cell impermeant stain that emits bright far-red fluorescence when bound to DNA

  • Cells that have lost plasma membrane integrity are stained within minutes, making this extremely useful stain to estimate live/dead cell ratios and to measure cytotoxicity in kinetic live cell assays
  • Suitable for staining nuclei in fixed cell preparations and tissue section
  • Formulated as room temperature-stable solution and comes in convenient dropper bottle: tip and drip 2 drops/mL to stain cells, then incubate 15 to 30 minutes in most cases
  • Exceptionally bright far-red fluorescence upon binding to DNA
  • Rapid staining of damaged, dead, or fixed cells without wash steps
  • Ready-to-use liquid formulation: no need to dilute, weigh, or pipet
  • High-affinity DNA stain that easily stains cells with compromised cell membranes, yet does not enter living cells
  • Fluorescence increases several hundred-fold on binding to dsDNA in cells with compromised cell membranes (dying or dead cells), in fixed cells, and tissue slices
  • Spectral properties, with excitation/emission at 642/661nm when bound to DNA, is excellent for standard Cy5/Far Red filters
  • Long-wavelength fluorescence of the dye is well separated from that of commonly used fluorophores, such as Hoechst, GFP, Alexa Fluor 488, rhodamine (TRITC), Texas Red and many RFP variants
  • Well-suited for tissue staining, as its signal is generally not obscured by tissue autofluorescence
  • May be added directly to cells in full media or buffer solutions
  • In most cases, staining intensity increases with time if cells are not washed prior to imaging.
  • Excited with a maximum at 642nm when bound to DNA, with an emission maximum at 661nm
  • It is detected through far-red filters

Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry of Cellular Processes, Fluorescence In Situ Hybridization, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, In Situ Hybridization (ISH), Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

TRUSTED_SUSTAINABILITY

Specifications

Color Far-red
Content And Storage 6 x 2.5 mL dropper bottles

Store at ≤ 25°C
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Flow Cytometer
Product Type Nucleic Acid Stain
Emission 661
Excitation Wavelength Range 642 nm
Form Liquid
Product Line NucRed, ReadyProbes
Quantity 6 Bottles kit
Sub Cellular Localization Nucleus
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.