missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nonyl Acridine Orange (Acridine Orange 10-Nonyl Bromide)

Product Code. 11573997
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11573997

Brand: Invitrogen™ A1372

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Well retained in mitochondria making it useful probe for mitochondria during isolation and after cell fusion

The mitochondrial uptake is reported to not to depend on membrane potential, unlike many other mitochondrial probes. It is toxic at high concentrations and apparently binds to cardiolipin in all mitochondria, regardless of their energetic state. This derivative has been used to analyze mitochondria by flow cytometry, to characterize multidrug resistance and to measure changes in mitochondrial mass during apoptosis.

Apoptosis, Cell Analysis, Cell Counting & Viability, Cell Counting, Viability, & Cryopreservation, Cell Culture, Cell Structure, Cell Viability, Proliferation & Function, DNA Fragmentation, Flow Cytometry, Flow Cytometry Apoptosis Assays, Mitochondria, Mitochondria Function, Mitochondrial Function, Mitochondrial Structure

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Mitochondria Probe
SubCellular Localization Mitochondria

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.