missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Newport Green™ DCF Diacetate, cell permeant

Product Code. 10696303
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10696303 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10696303 Supplier Invitrogen™ Supplier No. N7991
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Has moderate zinc-binding affinity (Kd for Zn2+ approximately 1μM) but is essentially insensitive to Ca2+.

  • Kd for Ca2+ >100μM), making this valuable probe for detecting Zn2+ influx into neurons through voltage- or glutamate-gated channels
  • When used alongside dyes with dual Ca2+ / Zn2+ sensitivity such as fura-2 and mag-fura-2, provides confirmation that changes in Zn2+ levels, and not Ca2+ or Mg2+ are being detected
  • The response to transition metals, including Zn, Mn, Fe, Co, Cu(I), Cu(II), Ni and Cd, has been characterized
  • Label (Ex/Em): Newport Green DCF (approximately 505/535nm)
  • Kd (Zn2+) (in buffer): approximately 1μM
  • Lyophilized product may be dissolved in DMSO for use
  • Product is typically loaded into cells by adding dissolved indicator to medium containing cells

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Zinc and Heavy Metal Ion Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope
Product Type DCF Diacetate
Dye Type Zinc Indicator
Product Line Newport Green
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.