missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MQAE (N-(Ethoxycarbonylmethyl)-6-Methoxyquinolinium Bromide)

Product Code. 11544746
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11544746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11544746 Supplier Invitrogen™ Supplier No. E3101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

6-methoxyquinolinium derivative and fluorescent indicator for intracellular Cl-

  • Detects ion via diffusion-limited collisional quenching
  • MQAE has greater sensitivity to Cl- and higher fluorescence quantum yield than SPQ

Anion Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Confocal Microscope, Flow Cytometer
Product Type MQAE
Dye Type Fluorescent Dye-Based
Quantity 100 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.