Learn More
Invitrogen™ Measure-IT™ High-Sensitivity Nitrite Assay Kit
Description
Measure-iT™ assay kits bring add-and-read convenience and high-throughput scalability to the quantitation of nitrite, thiol, and lead/cadmium. The Measure-iT™ High-Sensitivity Nitrite Assay Kit features solid reproducibility over a wide quantitation range, as well as excellent tolerance of common sample contaminants. The assay is linear from 20 to 500 pmoles of nitrite, making it up to 50 times more sensitive than colorimetric assays based on the Griess reagent. The kit includes a nitrite quantitation standard and a protocol that's optimized for microplate-based application.
For Research Use Only. Not for use in diagnostic procedures.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 1 Kit |
| For Use With (Application) | Nitrite Quantitation |
| Detection Method | Fluorescence |
| Product Line | Measure-IT |
| Product Type | Nitrite Assay Kit |
| Excitation Wavelength Range | 365 nm |
| Emission Wavelength Range | 450 nm |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.