missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mag-Fura-2, AM, cell permeant

Product Code. 11599076
Change view
Click to view available options
Quantity:
1mg
20 x 50 μg
Unit Size:
1000µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11599076 20 x 50 μg 1000µg
10673863 1mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11599076 Supplier Invitrogen™ Supplier No. M1292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Intracellular magnesium indicator that is ratiometric and UV light-excitable

  • Acetoxymethyl (AM) ester form
  • Useful for noninvasive intracellular loading

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Magnesium Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Magnesium Indicator
Dye Type Fluorescent Dye-Based
Quantity 20 x 50 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.