missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LysoSensor™ Yellow/Blue DND-160 - Special Packaging

Product Code. 11534986
Change view
Click to view available options
Quantity:
20 x 50 μL
Unit Size:
1000µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11534986 20 x 50 μL 1000µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11534986 Supplier Invitrogen™ Supplier No. L7545
  • ONLINE EXCLUSIVE PRICE
    £446.00
    £410.65 / 1000µL
    Save £35.35
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A ratiometric probe that can be used to measure the pH of acidic organelles, such as lysosomes

The LysoSensor dye produces blue fluorescence in neutral environments, but changes to yellow fluorescence in more acidic environments. The pKa of LysoSensor Yellow/Blue DND-160 is ∽4.2. Special packaging.

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Ionic Homeostasis & Signaling, Lysosomes, Organelle Tracing, pH Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow, Blue
Concentration 1 mM
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Product Line LysoSensor
Quantity 20 x 50 μL
Shipping Condition Room Temperature
Sub Cellular Localization Lysosomes
Emission Wavelength Range 329/440
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.