missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Sperm Viability Kit

Product Code. 10083162
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10083162 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10083162 Supplier Invitrogen™ Supplier No. L7011
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Assay for viability and fertilizing potential of sperm

The LIVE/DEAD Sperm Viability Kit provides a novel fluorescence-based assay for analyzing the viability and fertilizing potential of sperm by fluorescence microscopy or flow cytometry. Membrane-permeant SYBR™ 14 nucleic acid stain labels live sperm with green fluorescence, and membrane-impermeant propidium iodide labels the nucleic acids of membrane-compromised sperm with red fluorescence.

Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Flow Cytometry, Flow Cytometry Viability and Cytotoxicity Assays, Flow Cytometry of Cellular Processes

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Eukaryotic Cells, Sperm Cells
Color Green, Red
Content And Storage Store in freezer at -5°C to -30°C and protect from light.
Description LIVE/DEAD™ Sperm Viability Kit
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Sperm Viability Kit
Dye Type Other Label(s) or Dye(s)
Format Tube(s), Slide(s)
Product Line LIVE/DEAD
Quantity 1 kit
Shipping Condition Room Temperature
Emission Wavelength 516/617
Show More Show Less
What is the fluorescence emission maxima of SYBR 14 dye and propidium iodide?

When bound to DNA, the fluorescence emission maxima of these dyes are 516 nm and 617 nm, respectively. The spectra are available in Figure 1 of the Product Manual.

How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.