missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipopolysaccharides From Escherichia coli Serotype 055:B5, Alexa Fluor™ 568 Conjugate
Description
Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Product Type | Lipopolysaccharide Conjugate |
| Dye Type | Alexa Fluor Dyes |
| Form | Solid |
| Product Line | Alexa Fluor |
| Quantity | 100 μg |
| Shipping Condition | Room Temperature |
| Excitation/Emission | 578/603 |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?