missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipopolysaccharides From Escherichia coli Serotype 055:B5, Alexa Fluor™ 488 Conjugate

This bright, green-fluorescent conjugate can be used to follow lipopolysaccharide binding, transport and cell-internalization processes

Brand:  Invitrogen™ L23351

Code : 50

Additional Details : Weight : 0.00525kg

Product Code. 11534966

  • £266.00 / 100µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store in freezer (-5 to -30°C) and protect from light.
Lipopolysaccharide Conjugate
Solid
Room Temperature
Fluorescence
Alexa Fluor Dyes
Alexa Fluor™
495/519
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.