Learn More
Invitrogen™ Lipophilic Tracer Sampler Kit
For use with fluorescence microscopy
Brand: Invitrogen™ L7781
323.00 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
The Lipophilic Tracer Sampler kit contains 1mg samples of nine different membrane stains.
Cell Analysis, Cell Tracing & Tracking, General Cell Tracing, Microbial Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature and protect from light. | |
Fluorescent | |
Liphophilic Tracer |
Fluorescence Microscope | |
Room Temperature | |
Cell Membranes, Lipids, Plasma Membrane |
For Research Use Only. Not for use in diagnostic procedures.