Learn More
Invitrogen™ Image-IT™ LIVE Lysosomal and Nuclear Labeling Kit (Counterstains For GFP-Expressing Cells)
Description
The Image-iT LIVE Lysosomal and Nuclear Labeling Kit provides two stains - red-fluorescent LysoTracker™ Red DND-99 dye and blue-fluorescent Hoechst 33342 dye - for highly selective staining of lysosomes and the nucleus, respectively, in live green-fluorescent protein (GFP)-transfected cells. When used according to the sample protocol, cell-permeant LysoTracker Red DND-99 provides highly selective lysosomal staining with minimal background. Hoechst 33342 dye, a cell-permeant nucleic acid stain that is selective for DNA and spectrally similar to DAPI, is UV excitable and emits blue fluorescence when bound to DNA. This dye should not interfere with GFP fluorescence and is retained after fixation and permeabilization.
Cell Analysis, Cell Structure, Cell-Based Reporter Assays, Cellular Imaging, Enzyme & Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Lysosomes, Nucleus, Nucleoli & Nuclear Envelope, Proteins, Expression, Isolation & Analysis, Reporter Assays
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Color | Red, Blue |
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 1 Kit |
| Detection Method | Fluorescence |
| Product Line | Image-iT |
| Shipping Condition | Room Temperature |
| Label Type | Fluorescent Dye |
| Product Type | Membrane and Nuclear Labeling |
| SubCellular Localization | Lysosomes, Nucleus |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.