missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Escherichia coli (K-12 strain) BioParticles™, Alexa Fluor™ 594 conjugate

Product Code. 12080126
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 12080126

Brand: Invitrogen™ E23370

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The fluorescence of the dye conjugates is uniformly intense over the pH range from 3 to 10.

Conjugates covalently labeled with fluorophores (special care has been taken to remove free dye after conjugation).

  • Label (Ex/Em): Alexa Fluor594 (approx. 590/617nm)
  • Conjugates are provided as lyophilized powders
  • There are approximately 3 x 108 E. coli or S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
  • Conjugates can be reconstituted in buffer of choice for use in phagocytosis assays
  • Fluorescence of conjugates that are bound to surface of the cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
  • In addition to cellular applications, fluorescent conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
  • Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type E.coli Conjugate
Dye Type Alexa Fluor™ 594
Form Lyophilized Powder
pH 3 to 10
Product Line Alexa Fluor, BioParticles
Quantity 2 mg
Shipping Condition Room Temperature
Species E. coli
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.