missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Escherichia coli (K-12 strain) BioParticles™, Alexa Fluor™ 488 conjugate
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
- Conjugates covalently labeled with fluorophore (removed free dye after conjugation)
- Label (Ex/Em): Alexa Fluor488 (approx. 495/519nm)
- Particle: E. coli (K-12 strain)
- Conjugates are provided as lyophilized powders
- Approximately 3 x 108 E. coli particles perm g solid and approximately 2 x 107 zymosan particles permg solid
- Can be reconstituted in buffer of choice for use in phagocytosis assays
- Fluorescence of conjugates that are bound to surface of cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
- I addition to cellular applications, fluorescent conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
- Small particles may also be useful references for light scattering studies because their sizes and shapes differ in characteristic ways
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Cell Type | Mammalian Cell |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | E.coli Conjugate |
| Dye Type | Alexa Fluor™ 488 |
| Form | Solid |
| Product Line | Alexa Fluor, BioParticles |
| Quantity | 2 mg |
| Shipping Condition | Dry Ice |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction