Learn More
Invitrogen™ Molecular Probes™ HCS Mitotic Index Kit
Description
The HCS Mitotic Index Kit represents a powerful image-based assay for the identification of compounds that affect mitotic progression. The kit provides a primary antibody against Histone H3 (pS10) as a sensitive index of mitosis and a secondary antibody conjugated to the Alexa Fluor™ 488 dye. DAPI and the HCS NuclearMask™ Deep Red stain (H10294) provide two choices in the kit for DNA profiling and cell demarcation. This kit is amenable to combination with other measurements such as DNA profiling, general cytotoxicity, or immunocytochemical detection of choice targets. The kit contains sufficient reagents to perform two, 96-well plates.
Cell Analysis, Cell Cycle, Cell Proliferation, Cell Viability, Proliferation and Function, Cellular Imaging, High-Content Screening (HCS)
Order Info
Shipping Condition: Dry ice. The kit contains sufficient reagents to perform two 96-well plates.
Specifications
Specifications
| Quantity | 1 kit |
| Detection Method | Fluorescence |
| Product Type | Mitotic Index Kit |
| Conjugate | Alexa Fluor 488, DAPI, HCS NuclearMask Deep Red Stain |
| Format | 96-well Plate |
| Color | Blue, Far-Red, Green |
| Content And Storage | Store at ≤-20°C, desiccate, protect from light |
| For Use With (Equipment) | Fluorescence Microscope, High Content Instrument |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.