missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HCS DNA Damage Kit

Product Code. 10565854
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10565854 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10565854 Supplier Invitrogen™ Supplier No. H10292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Kit contains sufficient reagents for performing two plates in a 96-well plate format.

Detects DNA damage and cytotoxicity

The HCS DNA Damage kit simultaneously and quantitatively measures two important cell-health parameters: DNA damage and cytotoxicity. DNA damage is detected using an antibody against phosphorylated H2AX (Ser139) which is induced in response to double-strand break (DSB) formation. To detect cytotoxicity, the Image-iT™ DEAD™ Green is an impermeant dye in healthy cells that becomes permeant when the plasma membrane of cells is compromised. Both indicators are amenable to fixation and permeabilization, thus allowing for multiplexing with other biomarkers of interest. The blue-fluorescent nuclear segmentation tool Hoechst 33342 is also included and stains DNA in both live and dead cells.

Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Color Orange, Blue, Green
Content And Storage The kit contains sufficient reagents for performing 2 plates in a 96-well plate format.
  • Store at ≤-20°C, desiccate and protect from light.
  • Detection Method Fluorescence
    For Use With (Equipment) High Content Analysis Instrument
    Product Type DNA Damage kit
    Dye Type Alexa Fluor™ 555, Hoechst 33258
    Format 96-well plate
    Product Line Molecular Probes
    Quantity 1 kit
    Shipping Condition Dry Ice

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.