Learn More
Invitrogen™ Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging
Product Code. 11534866
Click to view available options
Quantity:
10 x 100 μg
Unit Size:
1000µg
For Research Use Only. All usage must comply with product instructions.
Product Code. 11534866
Brand: Invitrogen™ G36000
For Research Use Only. All usage must comply with product instructions.
Glutathiolation detection reagent
Biotinylated glutathione ethyl ester (BioGEE) is cell-permeant, biotinylated glutathione analog for detection of glutathiolation Under conditions of oxidative stress, cells may transiently incorporate glutathione into proteins. Stressed cells incubated in BioGEE will also incorporate this biotinylated glutathione derivative into proteins, facilitating the identification of oxidation-sensitive proteins.
  • Glutathiolation levels can be detecte with fluorescent streptavidin conjugate using either flow cytometry or fluorescence microscopy
  • Proteins glutathiolated with BioGEE can also be extracted and analyzed by mass spectrometry or by Western blotting methods in conjunction with fluorophore- or enzyme-labeled streptavidin conjugates
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Glutathione Detection, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 x 100 μg
Product Type Glutathione Reagent