missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fura Red™, AM, cell permeant

Product Code. 11508966
Change view
Click to view available options
Quantity:
10 x 50 μg
500 μg
Unit Size:
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11508966 10 x 50 μg 500µg
11598956 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11508966 Supplier Invitrogen™ Supplier No. F3021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

Visible light-excitable fura-2 analog that offers particular possibilities for ratiometric measurement of Ca2+ in single cells

  • Measure by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicator
  • Acetoxymethyl (AM) ester form
  • Useful for noninvasive intracellular loading

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Cell-Based Second Messenger Assays, Drug Discovery and Development, G-Protein Coupled Receptor Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Fura Red AM
Dye Type Fluorescent Dye-Based
Product Line Fura Red
Quantity 10 x 50 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.