missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fura dextran, Potassium Salt, 10,000 MW, Anionic

Product Code. 11544816
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 11544816

Brand: Invitrogen™ F3029

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Derivative of ratiometric, UV light-excitable fura-2 indicator coupled to water-soluble 10,000 MW dextran

  • Useful for intracellular loading by microinjection, infusion from patch pipet or uptake induced by Influx™ pinocytic cell-loading reagent (Cat. No. I-14402)
  • Fura dextran conjugates tend to remain in cytosol without compartmentalization or leakage and are less likely to bind to cellular proteins, making them useful for long-term Ca2+ measurements

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Potassium Assay
For Use With (Equipment) Fluorescence Microscope, Microphotometer, Flow Cytometer
Product Type Fura Dextran Potassium Salt
Dye Type Fluorescent Dye-Based
Quantity 5 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.