missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FluoZin™-3, Tetrapotassium Salt, cell impermeant
Click to view available options
Quantity:
500 μg
Unit Size:
100µg
Description
- Exhibits high Zn2+-binding affinity that is unperturbed by Ca2+ concentrations up to at least 1μM
- Exhibits >50-fold increase in fluorescence in response to saturating levels of Zn2+
- Although imaging applications predominate, cell-based microplate assays and flow cytometry protocols have also been developed
- Label (Ex/Em): FluoZin-3 (approximately 494/516nm)
- Kd (Zn2+) (in buffer): approximately 15nM
- Lyophilized product may be dissolved in distilled water or aqueous buffer for use
- Product is typically loaded into cellsusing microinjection or diffusion from patch pipet
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Zinc and Heavy Metal Ion Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Zinc Indicator |
| Dye Type | Zinc Indicator |
| Product Line | FluoZin |
| Quantity | 500 μg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction