missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoZin™-1, Tripotassium Salt, cell impermeant

Product Code. 11574806
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
This item is not returnable. View return policy

Product Code. 11574806

Brand: Invitrogen™ F24180

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed to detect Zn2+ concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists

  • Based on N-(2-methoxyphenyl)iminodiacetate chelator,designed for detection of Zn2+ in the 0-100μM range with minimal interfering Ca2+ sensitivity
  • Label (Ex/Em): FluoZin-1 (approximately 495/515nm)
  • Kd (Zn2+) (in buffer): approximately 8μM
  • Lyophilized product may be dissolved in distilled water or aqueous buffer for use
  • Product is typically loaded into cells using microinjection or diffusion from patch pipet

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation, Zinc and Heavy Metal Ion Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Zinc Indicator
Dye Type Zinc Indicator
Product Line FluoZin
Quantity 500 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.