missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoroMyelin™ Green Fluorescent Myelin Stain - Solution in Water

Product Code. 10513653
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10513653 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10513653 Supplier Invitrogen™ Supplier No. F34651
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

For use with fluorescence microscopy

The FluoroMyelin Green fluorescent myelin stain enables quick and selective labeling of myelin in brain cryosections in a single 20-minute labeling step plus washes. This stain can be used in conjunction with antibodies and other dyes, and with standard histochemical methods for cryosection material. Solution in water.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Neuronal Tracing

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Detection Method Fluorescence
Product Type Green Fluorescent Myelin Stain
Quantity 1 mL
Content And Storage Store at room temperature and protect from light.
Label or Dye FluoroMyelin™ Green
Shipping Condition Room Temperature
For Use With (Equipment) Fluorescence Microscope
Sub Cellular Localization Myelin
Excitation/Emission 479/598 nm
Product Line FluoroMyelin
Show More Show Less
I used one of your FluoroMyelin stains and noticed it stains cells other than glial cells. Is there something wrong with the product?

FluoroMyelin is a lipid stain, any lipid can be stained by it but there is a higher lipid content in myelin that it will stain much more intensely than other membranes.

Can the FluoroMyelin stains be used on neurons in culture?

We have only tested them on mouse brain cryosections. They might work on neurons in culture, but would likely stain the membrane and there would be some endocytosis of the stains. Myelin sheaths may show enhanced intensity.

Can the FluoroMyelin myelin stains be used on paraffin sections?

We have only tested them on mouse brain cryosections.

What are the FluoroMyelin myelin stains?

They are proprietary lipid stains that stain myelin, but are not neuron specific.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.