missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein Diacetate (FDA)

Product Code. 10694763
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10694763 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10694763 Supplier Invitrogen™ Supplier No. F1303

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Cell-permeant esterase substrate that can serve as viability probe

  • Measures both enzymatic activity required to activate its fluorescence and cell-membrane integrity, intracellular retention of their fluorescent product
  • Upon hydrolysis by intracellular esterases, AM ester yields fluorescein

Cell Adhesion, Cell Analysis, Cell Proliferation, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, Target-Based ADME/Tox Assays, pH Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Product Type Fluorescein Diacetate
Dye Type Classic Dyes
Quantity 1 g
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.