missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluo-5N, AM, cell permeant - Special Packaging
Click to view available options
Quantity:
10 x 50 μg
Unit Size:
500µg
Description
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+.
- Cells may be physically loaded with cell-impermeant salt forms of these indicators using patch pipet or microinjection or Influx™ pinocytotic cell-loading reagent
- Compatible with excitation at 488nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- BAPTA-based indicators bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
- Perturbations to calcium measurements caused by presence of the ions can be controlled using heavy metal-selective chelator
- Label (Ex/Em of Ca2+-bound form): Fluo-5N (494/516nm)
- Fluorescence intensity increase upon binding Ca2+: >100 fold
- Kd for Ca2+ in buffer: approximately 90μM
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager |
| Product Type | Calcium Indicator |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 10 x 50 μg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction