missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FilmTracer™ SYPRO™ Ruby Biofilm Matrix Stain

Product Code. 11515336
Click to view available options
Quantity:
200 mL
Unit Size:
200mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11515336

Brand: Invitrogen™ F10318

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

5 ingredients

Convenient, ready-to-use 1X concentration

FilmTracer SYPRO Ruby biofilm matrix stain has been used to stain matrices of biofilms. This stain was originally used as a gel stain that labels most classes of proteins, including glycoproteins, phosphoproteins, lipoproteins, calcium binding proteins, fibrillar proteins and other proteins that are difficult to stain.

  • Flow Cytometer Laser Lines: 488
  • Platfotm: Confocal Microscope, Fluorescent Imager, Microplate Reader, Fluorescence Microscope
  • Label or Dye: SYPRO™ Ruby

Analysis of Bacteria, Bacterial Staining & Detection, Cell Analysis, Microbiological Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Content And Storage 5 ingredients, store at room temperature - may be exposed to light for a short period of time
Excitation Wavelength Range 450⁄610
Dye Type SYPRO™ Ruby
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Fluorescent Imager, Microplate Reader
Quantity 200 mL
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line FilmTracer, Molecular Probes
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Biofilm Matrix Stain
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.